MilliporeSigma
  • Synthesis of novel series of malonamides derivatives via a five-component reaction.
Synthesis of novel series of malonamides derivatives via a five-component reaction.
Molecular diversity (2013-05-25)
Abbas Rahmati, Tahmineh Kenarkoohi, Mahdi Ahmadi-Varzaneh
ABSTRACT
A one-pot, five-component condensation reaction of isocyanide, Meldrum's acid, arylidene malononitrile, and two amine molecules in CH2Cl2 at ambient temperature to give malonamide derivatives is described.
Sigma-Aldrich
Malonamide, 97%