MilliporeSigma
  • Short-term treatment with cell wall degrading enzymes increases the activity of the inositol phospholipid kinases and the vanadate-sensitive ATPase of carrot cells.
Short-term treatment with cell wall degrading enzymes increases the activity of the inositol phospholipid kinases and the vanadate-sensitive ATPase of carrot cells.
Plant physiology (1990-12-01)
Q Chen, W F Boss
ABSTRACT
Treating carrot (Daucus carota L.) suspension culture cells with a mixture of cell wall degrading enzymes, Driselase, resulted in an increase in the percentage of [(3)H]phosphatidylinositol bisphosphate. Analysis of the lipid kinase activities in the isolated plasma membranes after whole cell treatment indicated that treatment with Driselase (2% weight/volume; the equivalent of 340 units per milliliter of hemicellulase and 400 units per milliliter of cellulase activity) or treatment with hemicellulase (31.7% weight/volume, 20.7 units per milliliter) resulted in an increase in the inositol phospholipid kinase activity. However, treatment with cellulase alone had no effect at 0.5% (weight/volume, 17.2 units per milliliter) or inhibited the kinase activity at 1% (weight/volume, 34.4 units per milliliter). The active stimulus in Driselase was heat sensitive. The plasma membrane vanadate-sensitive ATPase activity also increased when the cells were treated with Driselase. A time course study indicated that both the inositol phospholipid kinases and the plasma membrane vanadate-sensitive ATPase responded to as little as 5 seconds of treatment with 2% Driselase. However, at the lowest concentration of Driselase (0.04%, weight/volume) that resulted in an increase in inositol phospholipid kinase activity, the ATPase activity was not affected. Because inositol phospholipids have been shown to activate the vanadate-sensitive ATPase from plants (AR Memon, Q Chen, WF Boss [1989] Biochem Biophys Res Commun 162: 1295-1301), a stimulus-response pathway involving both the inositol phospholipid kinases and the plasma membrane vanadate-sensitive ATPase activity is discussed.
Sigma-Aldrich
Driselase from Basidiomycetes sp., powder, Protein ≥10 % by biuret