MilliporeSigma
  • Isolation of axial conformers of chloro- and bromocyclohexane in a pure state as inclusion complexes with 9,9'-bianthryl, and the discovery of a novel 1,3 diaxial Cl...H weak interaction.
Isolation of axial conformers of chloro- and bromocyclohexane in a pure state as inclusion complexes with 9,9'-bianthryl, and the discovery of a novel 1,3 diaxial Cl...H weak interaction.
Chemical communications (Cambridge, England) (2005-07-20)
Shinya Hirano, Shinji Toyota, Masako Kato, Fumio Toda
ABSTRACT
The axial conformers of chloro- and bromocyclohexane were isolated in a pure state as inclusion complexes with 9,9'-bianthryl, and a 1,3 diaxial Cl...H weak interaction was discovered by X-ray analysis of the axial conformer of chlorocyclohexane.
Sigma-Aldrich
Bromocyclohexane, 98%