MilliporeSigma
  • Isolation of flavonoids and a chalcone from Helichrysum odoratissimum and synthesis of helichrysetin.
Isolation of flavonoids and a chalcone from Helichrysum odoratissimum and synthesis of helichrysetin.
Journal of natural products (1989-05-01)
L Van Puyvelde, N De Kimpe, J Costa, V Munyjabo, S Nyirankuliza, E Hakizamungu, N Schamp
ABSTRACT
3,5-Dihydroxy-6,7,8-trimethoxyflavone, 3-O-methylquercetin, and helichrysetin were isolated from the flowers of the Rwandese medicinal plant, Helichrysum odoratissimum. Because of inconsistencies of the mp of the latter chalcone, a synthesis of helichrysetin was developed. 3-O-Methylquercetin was shown to be an active principle as it displayed antimicrobial activity.
Sigma-Aldrich
3-O-Methylquercetin, ≥97% (HPLC)