MilliporeSigma
  • Enhancing effects of captafol on the development of GST-P-positive liver cell foci in a medium-term bioassay, and protection by L-cysteine of the enhancement in rats.
Enhancing effects of captafol on the development of GST-P-positive liver cell foci in a medium-term bioassay, and protection by L-cysteine of the enhancement in rats.
Cancer letters (1997-01-01)
H C Kim, S W Cha, S W Song, C S Ha, S S Han, J K Roh, Y S Lee, F Furukawa, A Nishikawa, M Takahashi
ABSTRACT
The modifying effects of captafol and protective effects of L-cysteine on the development of glutathione S-transferase placental form-positive (GST-P +) foci of the liver and expression of proliferating cell nuclear antigen (PCNA) in the kidney were investigated in a medium-term bioassay using D-galactosamine (DGA) in rats. Male 6-week-old F344 rats were initially given a single i.p. injection (200 mg/kg) of diethylnitrosamine (DEN) and after 2 weeks on basal diet, received two i.p. injections of DGA (300 mg/kg) at the ends of weeks 2 and 5, and were fed a diet supplemented with test chemicals for weeks 3-8. Animals in group 1 were given 1500 ppm captafol in the diet, while group 2 received 1500 ppm captafol in diet as well as 1500 ppm L-cysteine in drinking water, animals in control group being given basal diet alone. Positive results regarding increased numbers and areas of GST-P + liver cell foci were obtained in rats treated with captafol alone. On the other hand, significant reduction by L-cysteine in the areas of GST-P + liver cell foci initiated by DEN and promoted by captafol was observed. In addition, the PCNA-labelling indices of renal tubule cells were elevated in rats treated with captafol alone and significantly reduced in rats treated simultaneously with L-cysteine. The protocol used in the present study therefore allowed the in vivo determination of promoting effects of captafol and inhibitory influence of L-cysteine by analyzing GST-P + foci in the livers as marker lesions, within a relatively short period of 8 weeks. Thus, this bioassay protocol could have applicability as a new in vivo assay system for the screening of hepatic carcinogenic or anti-carcinogenic agents.
Product Description
Supelco
Captafol, PESTANAL®, analytical standard