MilliporeSigma
  • Quantitative determination of norethisterone acetate in human plasma by capillary gas chromatography with mass-selective detection.
Quantitative determination of norethisterone acetate in human plasma by capillary gas chromatography with mass-selective detection.
Journal of chromatography. A (1996-10-25)
F Pommier, A Sioufi, J Godbillon
ABSTRACT
An analytical method for the determination of norethisterone acetate (NETA) in human plasma by capillary gas chromatography-mass-selective detection (GC-MS), with testosterone acetate as internal standard, was developed and validated. After addition of the internal standard, the compounds were extracted from plasma at basic pH into diethyl ether-dichloromethane (3:2, v/v), which was then evaporated to dryness. The compounds were converted into their pentafluoropropionyl derivatives which were determined by gas chromatography using a mass selective detector at m/z 486 for NETA and m/z 476 for the internal standard. Intra-day and inter-day accuracy and precision were found suitable over the range of concentrations between 0.10 to 10 ng/ml. The method was applied to clinical samples.
Product Description
Norethisterone acetate for system suitability, European Pharmacopoeia (EP) Reference Standard
Norethisterone acetate, European Pharmacopoeia (EP) Reference Standard