MilliporeSigma
  • Synthesis and structure-activity relationships of novel benzimidazole and imidazo[4,5-b]pyridine acid derivatives as thromboxane A2 receptor antagonists.
Synthesis and structure-activity relationships of novel benzimidazole and imidazo[4,5-b]pyridine acid derivatives as thromboxane A2 receptor antagonists.
Journal of medicinal chemistry (1993-04-30)
E Nicolaï, J Goyard, T Benchetrit, J M Teulon, F Caussade, A Virone, C Delchambre, A Cloarec
ABSTRACT
A series of 1-benzylbenzimidazole and 3-benzylimidazo[4,5-b]pyridine substituted in the 2-position by an alkanoic or mercaptoalkanoic acid chain was synthesized for evaluation as potential thromboxane A2/prostaglandin H2 (TXA2/PGH2) receptor antagonists. The affinity of each compound for washed human platelet TXA2/PGH2 receptors was determined by radioligand binding studies using [125I]PTA-OH. Structure-activity relationships led to the conclusions that 2-alkanoic acid derivatives were slightly more potent than 2-mercaptoalkanoic acids and that compounds possessing a 3,3-dimethylbutanoic acid in the 2-position were definitely the most potent with Ki values of 4-39 nM (11a, 11g-x, 37a, 37f-o, 23a-c). The replacement of this 3,3-dimethylbutanoic acid side chain by a shorter one led to a marked decrease of affinity (11b and 11c; Ki = 5600 and 1700 nM, respectively). Compounds of benzimidazole and imidazo[4,5-b]pyridine series displayed similar potencies (11q and 23c have Ki values of 6 and 7 nM, respectively). The interesting pharmacological profile of compound 23a (UP 116-77: 4-[3-[(4-chlorophenyl)methyl]-6-chloroimidazo[4,5-b]pyridin-2-yl]- 3,3-dimethylbutanoic acid) and its excellent tolerance led us to select this derivative for further development.
Sigma-Aldrich
4-Azabenzimidazole, 99%