MilliporeSigma
  • Enzyme catalysis in multiphase systems: direct determination of water distribution.
Enzyme catalysis in multiphase systems: direct determination of water distribution.
Bio/technology (Nature Publishing Company) (1993-11-01)
R A Stenning, E C Needs, B E Brooker, E N Vulfson
ABSTRACT
The distribution of water and the formation of microphases during the synthesis of octyl octanoate and the hydrolysis of triolein catalyzed by immobilized lipase have been directly monitored in real time using confocal laser microscopy.
Sigma-Aldrich
Octyl octanoate, ≥98%, FG