MilliporeSigma
Testicular hydrocele treated by bismuth phosphate injection.
International urology and nephrology (1988-01-01)
M Dimberg, H Norlén, L Zachrisson, H Lindholm
ABSTRACT
Thirty-eight testicular hydroceles were treated by injection with 0.2-0.4 g of bismuth phosphate. The follow-up was completed in 32 cases, 17 (53%) of which showed complete regression although one patient had to wait 24 months for full regression. Eight patients (25%) showed clinical improvement and one patient (3%) had residual scrotal tenderness after otherwise successful treatment. Six patients (19%) did not improve and were treated surgically. The method may present an alternative to surgery in elderly patients with testicular hydrocele.
Sigma-Aldrich
Bismuth(III) phosphate, 99.99%