MilliporeSigma
  • eIF4E-Overexpression imparts perillyl alcohol and rapamycin-mediated regulation of telomerase reverse transcriptase.
eIF4E-Overexpression imparts perillyl alcohol and rapamycin-mediated regulation of telomerase reverse transcriptase.
Experimental cell research (2013-06-12)
Tabetha Sundin, Dennis Peffley, Patricia Hentosh
ABSTRACT
Translation is mediated partly by regulation of free eukaryotic initiation factor 4E (eIF4E) levels through PI3K-Akt-mTOR signaling. Cancer cells treated with the plant-derived perillyl alcohol (POH) or the mechanistic target of rapamycin (mTOR) inhibitor rapamycin dephosphorylate eIF4E-binding protein (4E-BP1) and attenuate cap-dependent translation. We previously showed in cancer cell lines with elevated eIF4E that POH and rapamycin regulate telomerase activity through this pathway. Here, immortalized Chinese hamster ovary (CHO) control cells and CHO cells with forced eIF4E expression (rb4E) were used to elucidate eIF4E's role in telomerase regulation by POH and rapamycin. Despite 5-fold higher eIF4E amounts in rb4E, telomerase activity, telomerase reverse transcriptase (TERT) mRNA, and TERT protein were nearly equivalent in control and rb4E cells. In control cells, telomerase activity, TERT mRNA and protein levels were unaffected by either compound. In contrast, telomerase activity and TERT protein were both attenuated by either agent in rb4E cells, but without corresponding TERT mRNA decreases indicating a translational/post-translational process. S6K, Akt, and 4E-BP1 were modulated by mTOR mediators only in the presence of increased eIF4E. Thus, eIF4E-overexpression in rb4E cells enables inhibitory effects of POH and rapamycin on telomerase and TERT protein. Importantly, eIF4E-overexpression modifies cellular protein synthetic processes and gene regulation.
Product Description
Supelco
Sirolimus solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
(−)-Perillyl alcohol, analytical standard
Supelco
Rapamycin, VETRANAL®, analytical standard
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, 96%
Sigma-Aldrich
(S)-(−)-Perillyl alcohol, ≥95%, FG
Sigma-Aldrich
Rapamycin from Streptomyces hygroscopicus, ≥95% (HPLC), powder