MilliporeSigma
  • ELEMENTAL SELENIUM GENERATED BY THE PHOTOBLEACHING OF SELENOMEROCYANINE PHOTOSENSITIZERS FORMS CONJUGATES WITH SERUM MACROMOLECULES THAT ARE TOXIC TO TUMOR CELLS.
ELEMENTAL SELENIUM GENERATED BY THE PHOTOBLEACHING OF SELENOMEROCYANINE PHOTOSENSITIZERS FORMS CONJUGATES WITH SERUM MACROMOLECULES THAT ARE TOXIC TO TUMOR CELLS.
Phosphorus, sulfur, and silicon and the related elements (2006-02-24)
Fritz Sieber, Jean-Pierre Daziano, Wolfgang H H Günther, Marianne Krieg, Kiyoko Miyagi, Reynée W Sampson, Martin D Ostrowski, Gregory S Anderson, Ichiro Tsujino, Raymond J Bula
ABSTRACT
Elemental selenium generated by the photobleaching of selenomerocyanine dyes forms conjugates with serum albumin and serum lipoproteins that are toxic to leukemia and selected solid tumor cells but well tolerated by normal CD34-positive hematopoietic stem and progenitor cells. Serum albumin and lipoproteins act as Trojan horses that deliver the cytotoxic entity (elemental selenium) to tumor cells as part of a physiological process. They exploit the fact that many tumors have an increased demand for albumin and/or low-density lipoprotein. Se(0)-protein conjugates are more toxic than selenium dioxide, sodium selenite, selenomethionine, or selenocystine. They are only minimally affected by drug resistance mechanism, and they potentiate the cytotoxic effect of ionizing radiation and several standard chemotherapeutic agents. The cytotoxic mechanism of Se(0)-protein conjugates is not yet fully understood. Currently available data are consistent with the notion that Se(0)-protein conjugates act as air oxidation catalysts that cause a rapid depletion of intracellular glutathione and induce apoptosis. Drugs modeled after our Se(0)-protein conjugates may prove useful for the local and/or systemic therapy of cancer.
Sigma-Aldrich
2-Mercaptopyridine N-oxide, 99%
Supelco
N-Methyl-N-(trimethylsilyl)trifluoroacetamide, synthesis grade
Sigma-Aldrich
(tert-Butoxycarbonylmethylene)triphenylphosphorane, 98%
Sigma-Aldrich
Dimercaptomaleonitrile disodium salt hydrate, ≥95.0% (calc. based on dry substance, NT)
Sigma-Aldrich
2-Chloro-1,3,2-dioxaphospholane, 97%
Sigma-Aldrich
4-Amino-2,2,6,6-tetramethylpiperidine, 98%
Sigma-Aldrich
Hydrogen peroxide–Urea adduct, powder, 15-17% active oxygen basis
Sigma-Aldrich
Cesium carbonate, ReagentPlus®, 99%