MilliporeSigma
  • Metal-free, highly efficient organocatalytic amination of benzylic C-H bonds.
Metal-free, highly efficient organocatalytic amination of benzylic C-H bonds.
Chemical communications (Cambridge, England) (2013-03-29)
Qicai Xue, Jin Xie, Huamin Li, Yixiang Cheng, Chengjian Zhu
ABSTRACT
A new synthetic approach toward direct C-N bond formation through sp(3) C-H activation has been developed under metal-free conditions. Both primary and secondary benzylic C-H substrates could react smoothly with various amines to give only mono-amination products with good to excellent yields.
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, CP