MilliporeSigma
  • Stereodefined trisubstituted enolates as a unique entry to all-carbon quaternary stereogenic centers in acyclic systems.
Stereodefined trisubstituted enolates as a unique entry to all-carbon quaternary stereogenic centers in acyclic systems.
Nature protocols (2013-03-23)
Yury Minko, Morgane Pasco, Lukas Lercher, Ilan Marek
ABSTRACT
This protocol describes a new approach for the preparation of stereodefined trisubstituted chiral enolate species, avoiding conventional asymmetric enolization of carbonyl compounds. This protocol was developed as a single-flask synthetic sequence and therefore does not require isolation or purification of intermediate compounds. The sequence starts from a regioselective carbocupration reaction of readily accessible chiral ynamides; this is followed by oxidation of the generated vinylcuprate with a commonly available oxidizing reagent (tert-butyl hydroperoxide) in order to generate an enolate that completely retains its configuration. This synthetic protocol has been applied to the preparation of aldol and Mannich-type adducts. The procedure reported here requires a simple reaction setup commonly available in all synthetic laboratories and takes ∼6 h for completion and 2 h for isolation and purification. Final products are valuable diastereomerically and enantiomerically enriched building blocks for organic synthesis containing all-carbon quaternary stereocenters in acyclic systems.
Sigma-Aldrich
tert-Butyl hydroperoxide solution, CP
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in decane
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 70 wt. % in H2O
Sigma-Aldrich
tert-Butyl hydroperoxide solution, 5.0-6.0 M in nonane