MilliporeSigma
  • Asymmetric synthesis of L-carbidopa based on a highly enantioselective α-amination.
Asymmetric synthesis of L-carbidopa based on a highly enantioselective α-amination.
Organic letters (2013-03-13)
Àlex Pericas, Alexandr Shafir, Adelina Vallribera
ABSTRACT
A stereoselective synthesis of L-carbidopa in seven steps and 50% overall yield from commercial compounds is described. The key step involves a highly enantioselective α-amination reaction of an acyclic β-ketoester with di-tert-butyl azodicarboxylate induced by europium and (R,R)-diphenyl-pybox.
Product Description
Carbidopa, European Pharmacopoeia (EP) Reference Standard