MilliporeSigma
Implication of free cholesterol in LC-MS response enhancement.
Bioanalysis (2012-06-02)
Robert MacNeill, Amy Farside, Vishva Acharya, Ryan Stromeyer, Bryan Urbanowicz, Marc Moussallie
ABSTRACT
In the context of matrix effects in bioanalytical LC-MS/MS, a speculative link between free cholesterol, the recoveries of the compound from three common extraction procedures, and response enhancement was qualitatively investigated. Injections on-column of cholesterol both in solution and extracts, in conjunction with post-column infusion of three representative drugs, reveal a direct role in pronounced response enhancement for two out of three analytes, under one set of LC-MS/MS conditions, for the majority of typical plasma extraction procedures, where electrospray-based gaseous ion generation is used. Cholesterol has been shown to have a strong association with LC-MS/MS response enhancement and ideally should be monitored during method development, reinforcing the reasoning behind minimizing SPE elution volumes and avoiding less selective means of sample preparation.
Sigma-Aldrich
Rizatriptan benzoate salt, ≥98% (HPLC)