MilliporeSigma
  • Fabrication of large-area ordered and reproducible nanostructures for SERS biosensor application.
Fabrication of large-area ordered and reproducible nanostructures for SERS biosensor application.
The Analyst (2012-02-23)
Gobind Das, Niranjan Patra, Anisha Gopalakrishnan, Remo Proietti Zaccaria, Andrea Toma, Sanjay Thorat, Enzo Di Fabrizio, Alberto Diaspro, Marco Salerno
ABSTRACT
We propose a large-area SERS device with efficient fluorescence quenching capability. The substrate is based on anodic porous alumina templates with various pore size and wall thickness as small as 15 and 36 nm, respectively. The nano-patterned SERS substrate, with excellent control and reproducibility of plasmon-polaritons generation, shows very efficient enhanced Raman signal in the presence of intrinsically fluorescent molecules such as cresyl violet, rhodamine, and green fluorescent protein. This work demonstrates that, when the nanostructures are properly designed and fabricated, Raman and fluorescence spectroscopy can be used in combination in order to obtain complementary molecular informations. Theoretical simulation shows excellent agreement with the experimental findings. The enhancement factor is found to be 10(3)-10(4), with respect to flat gold surface when the molecules are supposed to be closely packed, with considerable fluorescence suppression, showing a promising disposable biosensor.
Sigma-Aldrich
Cresyl Violet acetate, certified by the BSC
Sigma-Aldrich
Cresyl Violet perchlorate