MilliporeSigma
Antipsychotic medication for early episode schizophrenia.
The Cochrane database of systematic reviews (2011-06-17)
John Bola, Dennis Kao, Haluk Soydan
ABSTRACT
Long-term treatment with antipsychotic medications in early episode schizophrenia spectrum disorders is common, but both short and long-term effects on the illness are unclear. There have been numerous suggestions that people with early episodes of schizophrenia appear to respond differently than those with multiple prior episodes. The number of episodes may moderate response to drug treatment. To assess the effects of antipsychotic medication treatment on people with early episode schizophrenia spectrum disorders. We searched the Cochrane Schizophrenia Group register (July 2007) as well as references of included studies. We contacted authors of studies for further data. Studies with a majority of first and second episode schizophrenia spectrum disorders comparing initial antipsychotic medication treatment with placebo, milieu, or psychosocial treatment. Working independently, we critically appraised records from 681studies, of which five studies met inclusion criteria. John Rathbone from the Schizophrenia Group supported us with the data extraction. We calculated risk ratios (RR) and their 95% confidence intervals (CI) where possible. For continuous data, we calculated mean difference (MD). We calculated numbers needed to treat/harm (NNT/NNH) where appropriate. Five studies with a combined N = 998 met inclusion criteria. Four studies (N = 724) provided leaving the study early data and results suggested that individuals treated with a typical antipsychotic medication are less likely to leave the study early than those treated with placebo (Chlorpromazine: 3 RCTs N = 353, RR 0.4 CI 0.3 to 0.5, NNT 3.2, Fluphenaxine: 1 RCT N = 240, RR 0.5 CI 0.3 to 0.8, NNT 5; Thioridazine: 1 RCT N = 236, RR 0.44 CI 0.3 to 0.7, NNT 4.3, Trifulperazine: 1 RCT N = 94, RR 0.96 CI 0.3 to 3.6). Two studies (Cole 1964; May 1976) contributed data to assessment of side effects and present a general pattern of more frequent side effects among individuals treated with typical antipsychotic medications compared to placebo. Rappaport 1978 suggested a higher rehospitalisation rate for those receiving chlorpromazine compared to placebo (N = 80, RR 2.29 CI 1.3 to 4.0, NNH 2.9). However, a higher attrition in the placebo group is likely to have introduced a survivor bias into this comparison, as this difference becomes non-significant in a sensitivity analysis on intent-to-treat participants (N = 127, RR 1.69 CI 0.9 to 3.0). One study (May 1976) contributes data to a comparison of trifluoperazine to psychotherapy on long-term health in favour of the trifluoperazine group (N = 92, MD 5.8 CI 1.6 to 0.0); however, data from this study are also likely to contain biases due to selection and attrition. One study (Mosher 1995) contributes data to a comparison of typical antipsychotic medication to psychosocial treatment on six-week outcome measures of global psychopathology (N = 89, MD 0.01 CI -0.6 to 0.6) and global improvement (N = 89, MD -0.03 CI -0.5 to 0.4), indicating no between-group differences. On the whole, there is very little useable data in the few studies meeting inclusion criteria. With only a few studies meeting inclusion criteria, and with limited useable data in these studies, it is not possible to arrive at definitive conclusions. The preliminary pattern of evidence suggests that people with early episode schizophrenia treated with typical antipsychotic medications are less likely to leave the study early, but more likely to experience medication-related side effects. Data are too sparse to assess the effects of antipsychotic medication on outcomes in early episode schizophrenia.
Product Description
Thioridazine for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Thioridazine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Fluphenazine dihydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®