MilliporeSigma
  • Effects of synthetic retinoid Am80 on iodide-induced autoimmune thyroiditis in nonobese diabetic mice.
Effects of synthetic retinoid Am80 on iodide-induced autoimmune thyroiditis in nonobese diabetic mice.
Cellular immunology (2011-05-24)
Kazuki Morohoshi, Katsumi Yoshida, Yoshinori Nakagawa, Saeko Hoshikawa, Hiroshi Ozaki, Yurie Takahashi, Sadayoshi Ito, Kouki Mori
ABSTRACT
We examined whether a synthetic retinoid Am80 prevented the development of autoimmune thyroiditis in iodide-treated nonobese diabetic mice, an animal model of Hashimoto's thyroiditis (HT). Am80 (0, 0.1 or 1 mg/kg/day) was orally administered in feed during the 8-week iodide treatment. While iodide ingestion effectively induced thyroiditis, Am80 administration failed to interfere with thyroiditis development and serum anti-thyroglobulin antibody levels regardless of the dose of the retinoid. Splenic T cell numbers, splenocyte proliferation and interferon-γ production were decreased in the Am80-treated mice. Our data suggest that Am80 is not a candidate for use in the prevention of HT.
Sigma-Aldrich
Tamibarotene, ≥98% (HPLC)