MilliporeSigma
Photochemistry of 5-aminoquinoline in protic and aprotic solvents.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2011-04-20)
Jagat P Bridhkoti, Hirdyesh Mishra, H C Joshi, Sanjay Pant
ABSTRACT
Photophysical properties of 5-aminoquinoline (5AQ) have been investigated in various non-polar and polar (protic and aprotic) solvents using steady state and time resolved fluorescence. In aprotic solvents, the spectral maxima depend on the polarity. However, in protic solvents both the fluorescence intensity as well decay time show decrease depending on the hydrogen bonding ability of the solvent. The results suggest that photochemistry 5AQ is quite sensitive towards the polarity as well as protic character of the solvent.
Sigma-Aldrich
5-Aminoquinoline, 97%