MilliporeSigma
  • Intravenous and sublingual buprenorphine in horses: pharmacokinetics and influence of sampling site.
Intravenous and sublingual buprenorphine in horses: pharmacokinetics and influence of sampling site.
Veterinary anaesthesia and analgesia (2011-04-20)
Kristen M Messenger, Jennifer L Davis, Douglas H LaFevers, Beth M Barlow, Lysa P Posner
ABSTRACT
To describe the pharmacokinetics and adverse effects of intravenous (IV) and sublingual (SL) buprenorphine in horses, and to determine the effect of sampling site on plasma concentrations after SL administration. Randomized crossover experiment; prospective study. Eleven healthy adult horses between 6 and 20 years of age and weighing 487-592 kg. In the first phase; buprenorphine was administered as a single IV or SL dose (0.006 mg kg(-1)) and pharmacokinetic parameters were determined for each route of administration using a noncompartmental model. In the second phase; the jugular and lateral thoracic veins were catheterized for simultaneous venous blood sampling, following a dose of 0.006 mg kg(-1) SL buprenorphine. For both phases, plasma buprenorphine concentrations were measured using ultra-performance liquid chromatography with mass spectrometry. At each sampling period, horses were assessed for behavioral excitement and gastrointestinal motility. Following IV administration, buprenorphine mean ± SD half-life was 5.79 ± 1.09 hours. Systemic clearance (Cl) following IV administration was 6.13 ± 0.86 mL kg(-1) minute(-1) and volume of distribution at steady-state was 3.16 ± 0.65 L kg(-1). Following IV administration, horses showed signs of excitement. Gastrointestinal sounds were decreased following both routes of administration; however, none of the horses exhibited signs of colic. There was a significant discrepancy between plasma buprenorphine concentrations measured in the jugular vein versus the lateral thoracic vein following phase 2, thus pharmacokinetic parameters following SL buprenorphine are not reported. Buprenorphine has a long plasma half-life and results in plasma concentrations that are consistent with analgesia in other species for up to 4 hours following IV administration of this dose in horses. While buprenorphine is absorbed into the circulation following SL administration, jugular venous sampling gave a false measurement of the quantity absorbed and should not be used to study the uptake from SL administration.
Product Description
Supelco
Norbuprenorphine solution, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Norbuprenorphine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®