MilliporeSigma
  • Kinetics of 3-chlorotyrosine formation and loss due to hypochlorous acid and chloramines.
Kinetics of 3-chlorotyrosine formation and loss due to hypochlorous acid and chloramines.
Chemical research in toxicology (2011-02-16)
Matthew P Curtis, Andrew J Hicks, Jonathan W Neidigh
ABSTRACT
The persistent activation of innate immune cells in chronic inflammation is gaining recognition as a contributing factor in a number of human diseases. A distinguishing feature of activated leukocytes at sites of inflammation is their production of reactive species such as hypochlorous acid (HOCl). Investigating the role of reactive molecules such as HOCl in inflammation and human disease requires appropriate biomarkers. The preferred biomarker for HOCl, and by extension its synthesizing enzyme myeloperoxidase, is 3-chlorotyrosine. 3-Chlorotyrosine is a chemically stable product formed when HOCl, or an HOCl-generated chloramine, reacts with the tyrosine side chain and is readily measured by sensitive mass spectrometry methods. However, Whiteman and Spencer ((2008) Biochem. Biophys. Res. Commun., 371, 50 - 53.) noted that 3-chlorotyrosine is degraded by HOCl, calling into question its use as a biomarker. The kinetic rate constants for the reaction of 3-chlorotyrosine with HOCl, histidine chloramine, or lysine chloramine to form 3,5-dichlorotyrosine are reported. The kinetics of tyrosine chlorination in the context of a peptide with a nearby lysine residue was also determined and further supports the role of chloramines in the chlorination of protein-bound tyrosine residues. The likelihood of free and protein-bound 3,5-dichlorotyrosine occurring in vivo, given the reported rate constants, is discussed.
Sigma-Aldrich
3-Chloro-L-tyrosine, 97%