MilliporeSigma
  • The effects of nicergoline on corneal nerve regeneration in rat corneas after photorefractive keratectomy.
The effects of nicergoline on corneal nerve regeneration in rat corneas after photorefractive keratectomy.
Current eye research (2010-12-23)
Su-Young Kim, Jiwook Yang, Young-Chun Lee
ABSTRACT
We wanted to investigate the effect of nicergoline on corneal nerve regeneration in rat corneas after photorefractive keratectomy (PRK). Twenty Sprague-Dawley male rats were divided into two groups, the control group and the group that had been treated with nicergoline for 4 weeks. Corneal wound healing was evaluated by fluorescein staining after PRK. Immunofluorescent staining was performed in the rat corneas at 1 month after PRK with monoclonal antibodies for class III β-tubulin, calcitonin gene-related peptide (CGRP), and substance P (SP). The stained nerve areas were calculated using an image-analysis program. The corneal wound healing rate was not significantly different between the nicergoline-treated rats and the control rats after PRK. At 1 month after PRK, the tubulin-positive, substance P-positive, and CGRP-positive nerve areas were significantly greater in the treatment group than those in the control group. Nicergoline treatment increased the corneal nerve area in the rats after they had undergone PRK. Nicergoline may help patients who have a decreased corneal sense, such as those with neurotrophic keratopathy and those patients after they undergo refractive surgery.
Product Description
Nicergoline for system suitability, European Pharmacopoeia (EP) Reference Standard
Supelco
Nicergoline, analytical standard, for drug analysis
Nicergoline for peak identification, European Pharmacopoeia (EP) Reference Standard