MilliporeSigma
Zinc signals promote IL-2-dependent proliferation of T cells.
European journal of immunology (2010-03-05)
Jennifer Kaltenberg, Laura M Plum, Julia L Ober-Blöbaum, Andrea Hönscheid, Lothar Rink, Hajo Haase
ABSTRACT
Zinc signals, i.e. a change of the intracellular concentration of free zinc ions in response to receptor stimulation, are involved in signal transduction in several immune cells. Here, the role of zinc signals in T-cell activation by IL-2 was investigated in the murine cytotoxic T-cell line CTLL-2 and in primary human T cells. Measurements with the fluorescent dyes FluoZin-3 and Zinquin showed that zinc is released from lysosomes into the cytosol in response to stimulation of the IL-2-receptor. Activation of the ERK-pathway was blocked by chelation of free zinc with N,N,N',N'-tetrakis-2(pyridyl-methyl)ethylenediamine, whereas zinc was not required for STAT5 phosphorylation. In addition, the key signaling molecules MEK and ERK were activated in response to elevated free intracellular zinc, induced by incubation with zinc and the ionophore pyrithione. Downstream of ERK activation, ERK-specific gene expression of c-fos and IL-2-induced proliferation was found to depend on zinc. Further experiments indicated that inhibition of MEK and ERK-dephosphorylating protein phosphatases is the molecular mechanism for the influence of zinc on this pathway. In conclusion, an increase of cytoplasmic free zinc is required for IL-2-induced ERK signaling and proliferation of T cells.
Sigma-Aldrich
Zinquin, ≥95% (HPLC), solid