MilliporeSigma
  • Development and validation of a dried blood spot-LC-APCI-MS assay for estimation of canrenone in paediatric samples.
Development and validation of a dried blood spot-LC-APCI-MS assay for estimation of canrenone in paediatric samples.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2010-02-16)
Maysa Faisal Suyagh, Prashant Laxman Kole, Jeff Millership, Paul Collier, Henry Halliday, James C McElnay
ABSTRACT
A selective and sensitive liquid chromatography (LC)-atmospheric pressure chemical ionisation (APCI)-mass spectroscopic (MS) assay of canrenone has been developed and validated employing Dried Blood Spots (DBS) as the sample collection medium. DBS samples were prepared by applying 30 microl of spiked whole blood onto Guthrie cards. A 6mm disc was punched from the each DBS and extracted with 2 ml of methanolic solution of 17alpha-methyltestosterone (Internal Standard). The methanolic extract was evaporated to dryness and reconstituted in acetonitrile:water (1:9, v/v). The reconstituted solution was further subjected to solid phase extraction using HLB cartridges. Chromatographic separation was achieved using Waters Sunfire C18 reversed-phase column using isocratic elution, followed by a high organic wash to clear late eluting/highly retained components. The mobile phase consisted of methanol:water (60:40, v/v) pumped at a flow rate of 0.3 ml/min. LC-APCI-MS detection was performed in the selected-ion monitoring (SIM) mode using target ions at m/z 341.1 and 303.3 for canrenone and internal standard respectively. The selectivity of the method was established by analysing DBS samples from 6 different sources (individuals). The calibration curve for canrenone was found to be linear over 25-1000 ng/ml (r>0.994). Accuracy (% RE) and precision (% CV) values for within and between day were <20% at the lower limit of quantification (LLQC) and <15% at all other concentrations tested. The LLOQ of the method was validated at 25 ng/ml. Clinical validation of the method was achieved by employing the validated method for analysis of 160 DBS samples from 37 neonatal and paediatric patients.
Product Description
Canrenone, European Pharmacopoeia (EP) Reference Standard