MilliporeSigma
  • Bioreduction of alpha-chloroacetophenone by whole cells of marine fungi.
Bioreduction of alpha-chloroacetophenone by whole cells of marine fungi.
Biotechnology letters (2009-06-06)
Lenilson C Rocha, Hercules V Ferreira, Eli F Pimenta, Roberto G S Berlinck, Mirna H R Seleghim, Darci C D Javaroti, Lara D Sette, Rafaella C Bonugli, André L M Porto
ABSTRACT
The asymmetric reduction of 2-chloro-1-phenylethanone (1) by seven strains of marine fungi was evaluated and afforded (S)-(-)-2-chloro-1-phenylethanol with, in the best case, an enantiomeric excess of 50% and an isolated yield of 60%. The ability of marine fungi to catalyse the reduction was directly dependent on growth in artificial sea water-based medium containing a high concentration of Cl(-) (1.2 M). When fungi were grown in the absence of artificial sea water, no reduction of 1 by whole cells was observed. The biocatalytic reduction of 1 was more efficient at neutral rather than acidic pH values and in the absence of glucose as co-substrate.
Sigma-Aldrich
2-Chloroacetophenone, 98%
Sigma-Aldrich
2-Chloroacetophenone, purum, ≥98.0% (GC)