MilliporeSigma
  • Stimulation of intrachromosomal homologous recombination in mammalian cells by an inhibitor of poly(ADP-ribosylation).
Stimulation of intrachromosomal homologous recombination in mammalian cells by an inhibitor of poly(ADP-ribosylation).
Nucleic acids research (1991-11-11)
A S Waldman, B C Waldman
ABSTRACT
We determined the effect of 3-methoxybenzamide (3-MB), a competitive inhibitor of poly(ADP-ribose) polymerase (E.C. 2.4.2.30), on intrachromosomal homologous recombination in mouse Ltk- cells. We used a cell line that contained in its genome two defective Herpes thymidine kinase (tk) genes as closely linked direct repeats. Intrachromosomal homologous recombination events were monitored by selecting for tk-positive segregants that arose during propagation of the cells and recombination rates were determined by fluctuation analysis. We found that growth of cells in the continuous presence of 2mM 3-MB increased intrachromosomal recombination between 3 and 4-fold. Growth of cells in the presence of 2mM m-anisic acid, a non-inhibitory analog of 3-MB, had no effect on intrachromosomal recombination rates. Additionally, we found that 3-MB increased both gene conversions and crossovers to similar extents, adding to the evidence that these two types of intrachromosomal rearrangements share a common pathway. These findings contrast with our previous studies [Waldman, B.C. and Waldman, A.S. (1990) Nucleic Acids Res., 18, 5981-5988] in which we determined that 3-MB inhibits illegitimate recombination and has no effect on extrachromosomal homologous recombination in mouse Ltk- cells. An hypothesis is offered that explains the influence of 3-MB on different recombination pathways in mammalian cells in terms of the role that poly(ADP-ribosylation) plays in DNA break-repair.
Sigma-Aldrich
3-Methoxybenzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
3-Methoxybenzamide, 97%