MilliporeSigma
  • Selected regiocontrolled transformations applied to the synthesis of (1S)-cis-chrysanthemic acid from (1S)-3,4-epoxy-2,2,5,5-tetramethylcyclohexanol.
Selected regiocontrolled transformations applied to the synthesis of (1S)-cis-chrysanthemic acid from (1S)-3,4-epoxy-2,2,5,5-tetramethylcyclohexanol.
Chemical communications (Cambridge, England) (2008-10-03)
Alain Krief, Humaira Y Gondal, Adrian Kremer
ABSTRACT
(1S)-cis-Chrysanthemic acid has been prepared in a few steps with complete control of the relative and absolute stereochemistry using regiocontrolled epoxide ring opening, diol mono-oxidation and cyclopropanation.
Sigma-Aldrich
(+)-trans-Chrysanthemic acid, ≥97.0% (GC)