MilliporeSigma
  • Solvation of phenylglycine- and leucine-derived chiral stationary phases: molecular dynamics simulation study.
Solvation of phenylglycine- and leucine-derived chiral stationary phases: molecular dynamics simulation study.
The journal of physical chemistry. B (2008-09-25)
Sorin Nita, Natalie M Cann
ABSTRACT
A theoretical study of the solvation of ( R)- N-(3,5-dinitrobenzoyl)phenylglycine- and ( R)- N-(3,5-dinitrobenzoyl)leucine-derived chiral stationary phases (CSPs) is presented. Semiflexible models of the chiral selectors are prepared from B3LYP/6-311G** calculations, and these are used in the molecular dynamics simulations of the corresponding interface. The chiral interface is examined for four solvents: 100% hexane, 90:10 hexane:2-propanol, 80:20 hexane:2-propanol, and 100% 2-propanol. Despite the similarities between phenylglycine and leucine, the interfaces are distinct both in terms of the selector orientations at the surface and in the number of hydrogen bonds formed with 2-propanol. We also find that an increase in alcohol concentration alters the preferred orientations of the selectors.
Sigma-Aldrich
(R)-(−)-N-(3,5-Dinitrobenzoyl)-α-phenylglycine, 99%