MilliporeSigma
  • Intense blue up-conversion luminescence in Tm3+/Yb3+ codoped oxyfluoride glass-ceramics containing beta-PbF2 nanocrystals.
Intense blue up-conversion luminescence in Tm3+/Yb3+ codoped oxyfluoride glass-ceramics containing beta-PbF2 nanocrystals.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2005-08-13)
Junjie Zhang, Zhongchao Duan, Dongbing He, Shixun Dai, Liyan Zhang, Lili Hu
ABSTRACT
Up-conversion luminescence properties of a Tm3+/Yb3+ codoped oxyfluoride glass-ceramics under 980 nm excitation are investigated. Intense blue emission centered at 476 nm, corresponding to 1G4-->3H6 transitions of Tm3+ was simultaneously observed in the transparent oxyfluoride glass ceramics at room temperature. The intensity of the blue up-conversion luminescence in a 1 mol% YbF3-containing glass-ceramic was found to be about 40 times stronger than that in the precursor oxyfluoride glass. The reason for the intense Tm3+ up-conversion luminescence in the oxyfluoride glass-ceramics is discussed. The dependence of up-conversion intensities on excitation power and possible up-conversion mechanism are also evaluated.
Sigma-Aldrich
Lead(II) fluoride, 99.99% trace metals basis
Sigma-Aldrich
Lead(II) fluoride, powder, ≥99%