MilliporeSigma
  • Linking solution chemistry to crystal nucleation: the case of tetrolic acid.
Linking solution chemistry to crystal nucleation: the case of tetrolic acid.
Chemical communications (Cambridge, England) (2005-03-17)
S Parveen, R J Davey, G Dent, R G Pritchard
ABSTRACT
The application of FTIR spectroscopy to concentrated solutions of tetrolic acid shows, for the first time, a direct relationship between molecular self association in solution and H-bonded motifs in the subsequently crystallised solid phases.
Sigma-Aldrich
2-Butynoic acid, 98%