MilliporeSigma
Four new triterpenoids from Maytenus ilicifolia.
Journal of natural products (2004-03-27)
Ayumi Ohsaki, Yoji Imai, Meiko Naruse, Shin-Ichi Ayabe, Kanki Komiyama, Junko Takashima
ABSTRACT
Four new triterpenoids with various skeletons, maytefolins A-C (1-3) and uvaol-3-caffeate (4), were isolated from the leaves of a Brazilian medicinal plant, Maytenus ilicifolia, together with five known triterpenoids. Of these triterpenoids only erythrodiol exhibited significant cytotoxicity against KB/S, KB/VJ300, and KU 19-20 cells.
Product Description
Supelco
Erythrodiol, analytical standard