MilliporeSigma
Fragrance material review on linalyl butyrate.
Food and chemical toxicology : an international journal published for the British Industrial Biological Research Association (2003-06-14)
C S Letizia, J Cocchiara, J Lalko, A M Api
ABSTRACT
A toxicologic and dermatologic review of linalyl butyrate when used as a fragrance ingredient is presented.
Sigma-Aldrich
Linalyl butyrate, ≥95%, FG