MilliporeSigma
  • Differential evolution: crystal structure determination of a triclinic polymorph of adipamide from powder diffraction data.
Differential evolution: crystal structure determination of a triclinic polymorph of adipamide from powder diffraction data.
Chemical communications (Cambridge, England) (2002-07-19)
Colin C Seaton, Maryjane Tremayne
ABSTRACT
The crystal structure of a previously unknown triclinic polymorph of adipamide has been solved from laboratory X-ray powder diffraction data using a new direct space global optimisation method based on differential evolution.
Sigma-Aldrich
Adipamide, 97%