MilliporeSigma
  • Protective activity of aromatic amines and imines against oxidative nerve cell death.
Protective activity of aromatic amines and imines against oxidative nerve cell death.
Biological chemistry (2002-01-05)
B Moosmann, T Skutella, K Beyer, C Behl
ABSTRACT
Oxidative stress is a widespread phenomenon in the pathology of neurodegenerative diseases such as Alzheimer's disease, Parkinson's disease, and amyotrophic lateral sclerosis. Neuronal cell death due to oxidative stress may causally contribute to the pathogeneses of these diseases. Therefore, neuroprotective antioxidants are considered to be a promising approach to slow down disease progression. We have investigated different aromatic amine and imine compounds for neuroprotective antioxidant functions in cell culture, and found that these compounds possess excellent cytoprotective potential in diverse paradigms of oxidative neuronal cell death, including clonal cell lines, primary cerebellar neurons, and organotypic hippocampal slice cultures. Aromatic amines and imines are effective against oxidative glutamate toxicity, glutathione depletion, and hydrogen peroxide toxicity. Their mode of action as direct antioxidants was experimentally confirmed by electron spin resonance spectroscopy, cell-free brain lipid peroxidation assays, and intracellular peroxide measurements. With half-maximal effective concentrations of 20-75 nM in different neuroprotection experiments, the aromatic imines phenothiazine, phenoxazine, and iminostilbene proved to be about two orders of magnitude more effective than common phenolic antioxidants. This remarkable efficacy could be directly correlated to calculated properties of the compounds by means of a novel, quantitative structure-activity relationship model. We conclude that bridged bisarylimines with a single free NH-bond, such as iminostilbene, are superior neuroprotective antioxidants, and may be promising lead structures for rational drug development.
Sigma-Aldrich
5H-Dibenz[b,f]azepine, 97%
Supelco
Iminostilbene, Pharmaceutical Secondary Standard; Certified Reference Material