MilliporeSigma
  • Kinetic analysis of enantiomers of threo-methylphenidate and its metabolite in two healthy subjects after oral administration as determined by a gas chromatographic-mass spectrometric method.
Kinetic analysis of enantiomers of threo-methylphenidate and its metabolite in two healthy subjects after oral administration as determined by a gas chromatographic-mass spectrometric method.
Journal of pharmaceutical sciences (1990-06-01)
T Aoyama, H Kotaki, Y Honda, F Nakagawa
ABSTRACT
A gas chromatographic-mass spectrometric method was developed for the stereoselective quantification of threo-methylphenidate (MPD) and its metabolite, ritalinic acid (RA), in plasma or urine. The plasma concentrations of (+)-MPD after oral administration of two 10-mg conventional tablets containing racemic MPD.HCl or of 20-mg of racemic MPD.HCl crystals to two healthy subjects were much higher than those of the (-)-isomer. The plasma concentrations of the metabolite, (-)-RA, were higher than that of the (+)-isomer during the first 4 h after administration of racemic MPD.HCl in both tablet and crystal forms. Although in urine both (+)- and (-)-RA were largely excreted in 48 h (37 and 40% of the dose, respectively), the percentage excretion of (-)-RA during the first 3-4 h was approximately twice that of the (+)-isomer. These results suggest that one reason for the difference in the plasma levels between (+)- and (-)-MPD may be due to differences in their rates of metabolism. Pharmacokinetic parameters of (+)-MPD after administration of 10 mg of (+)-MPD.HCl crystals were almost the same as those after administration of racemic MPD.HCl crystals. The AUC infinity 0 of (-)-MPD after administration of 10 mg of (-)-MPD.HCl crystals was smaller than that after administration of racemic MPD.HCl crystals.
Sigma-Aldrich
Ritalinic acid, 99% (CP)
Supelco
Ritalinic acid hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®