MilliporeSigma
  • Inhibition of protein synthesis on the ribosome by tildipirosin compared with other veterinary macrolides.
Inhibition of protein synthesis on the ribosome by tildipirosin compared with other veterinary macrolides.
Antimicrobial agents and chemotherapy (2012-08-29)
Niels Møller Andersen, Jacob Poehlsgaard, Ralf Warrass, Stephen Douthwaite
ABSTRACT
Tildipirosin is a 16-membered-ring macrolide developed to treat bacterial pathogens, including Mannheimia haemolytica and Pasteurella multocida, that cause respiratory tract infections in cattle and swine. Here we evaluated the efficacy of tildipirosin at inhibiting protein synthesis on the ribosome (50% inhibitory concentration [IC(50)], 0.23 ± 0.01 μM) and compared it with the established veterinary macrolides tylosin, tilmicosin, and tulathromycin. Mutation and methylation at key rRNA nucleotides revealed differences in the interactions of these macrolides within their common ribosomal binding site.
Product Description
Supelco
Tilmicosin, VETRANAL®, analytical standard, mix of isomers