MilliporeSigma
  • Encapsulation of the ethylene inhibitor 1-Methylcyclopropene by cucurbit[6]uril.
Encapsulation of the ethylene inhibitor 1-Methylcyclopropene by cucurbit[6]uril.
Journal of agricultural and food chemistry (2011-09-08)
Quan Zhang, Zeng Zhen, Hong Jiang, Xue-Gang Li, Jun-An Liu
ABSTRACT
1-Methylcyclopropene (1-MCP) is an excellent safe and commercially available ethylene antagonist for the preservation of horticultural products. However 1-MCP has to be stored in absorbents due to its gaseous and unstable characteristics. In this paper cucurbit[6]uril (CB[6]) was used as the absorbent to encapsulate 1-MCP, and the resultant inclusion complex was characterized by IR, powder X-ray diffraction, thermal analysis, and fluorescent spectra. The effects of encapsulation conditions on the formation of inclusion complex were also investigated. The amount of 1-MCP encapsulated by CB[6] was about 4.5% by weight when the initial concentration of 1-MCP, encapsulation temperature, CB[6] concentration, and encapsulation time were set at 75 mL/L, 20 °C, 30 mM, and 8 h, respectively. Furthermore, the release of 1-MCP from the complex can be realized with different solutions such as sodium bicarbonate, benzoic acid, and distilled water. CB[6] can be used as an excellent absorbent for encapsulation of 1-MCP.
Sigma-Aldrich
Cucurbit[6]uril hydrate, contains acid of crystalization