MilliporeSigma
  • Myeloperoxidase-mediated bioactivation of 5-hydroxythiabendazole: a possible mechanism of thiabendazole toxicity.
Myeloperoxidase-mediated bioactivation of 5-hydroxythiabendazole: a possible mechanism of thiabendazole toxicity.
Toxicology in vitro : an international journal published in association with BIBRA (2011-04-26)
Joseph D Jamieson, Evan B Smith, Deepak K Dalvie, Gregory J Stevens, Gina M Yanochko
ABSTRACT
Thiabendazole (TBZ), an antihelminthic and antifungal agent, is associated with a host of adverse effects including nephrotoxicity, hepatotoxicity, and teratogenicity. Bioactivation of the primary metabolite of TBZ, 5-hydroxythiabendazole, has been proposed to yield a reactive intermediate. Here we show that this reactive intermediate can be catalyzed by myeloperoxidase (MPO), a neutrophil-bourne peroxidase. Using a cell viability endpoint, we examined the toxicity of TBZ, 5OH-TBZ, and MPO-generated metabolites in cell-based models including primary rat proximal tubule epithelial cells, NRK-52E rat proximal tubule cells, and H9C2 rat myocardial cells. Timecourse experiments with MPO showed complete turnover of 5OH-TBZ within 15 min and a dramatic leftward shift in dose-response curves after 12h. After a 24h exposure in vitro, the LC(50) of this reactive intermediate was 23.3 ± 0.2 μM reduced from greater than 200 μM from 5OH-TBZ alone, an approximately 10-fold decrease. LC(50) values were equal in all cell types used. Comparison of lactate dehydrogenase leakage and caspase 3/7 activity revealed that cell death caused by the reactive intermediate is primarily associated with necrosis rather than apoptosis. This toxicity can be completely rescued via incubation with rutin, an inhibitor of MPO. These results suggest that MPO-mediated biotransformation of 5OH-TBZ yields a reactive intermediate which may play a role in TBZ-induced toxicity.
Product Description
Supelco
5-Hydroxythiabendazole, PESTANAL®, analytical standard