MilliporeSigma
  • Occurrence and fate of the human pharmaceutical metabolite ritalinic acid in the aquatic system.
Occurrence and fate of the human pharmaceutical metabolite ritalinic acid in the aquatic system.
Chemosphere (2010-10-12)
Marion Letzel, Klaus Weiss, Walter Schüssler, Manfred Sengl
ABSTRACT
To investigate the occurrence and fate of ritalinic acid - the main human metabolite of the psychostimulant drug methylphenidate - in the aquatic environment, a HPLC-electrospray-MS/MS method for the quantification of ritalinic acid in wastewater, surface water and bank filtrate was developed. Carbamazepine known as very stable in the aquatic environment was analyzed as anthropogenic marker in parallel. Furthermore, the removal of ritalinic acid was studied in a sewage treatment plant using an activated sludge system during a field study and in lab-scale plants. In good agreement between lab-scale and field studies a low removal rate of 13% and 23%, respectively, was determined. As a consequence, the concentration of ritalinic acid in the wastewater effluents were in the range of <50-170 ngL(-1) which corresponds to a mean specific load per capita of 17.7 μgd(-1). Ritalinic acid has further been detected in German rivers at concentrations of 4-23 ngL(-1) and in bank filtrate samples in 100-850 m distance from the river up to 5 ngL(-1) demonstrating the widespread occurrence of this stable metabolite in the aquatic environment. A comparison to available sales data shows that a significant amount of methylphenidate applied can be found in waters as ritalinic acid.
Product Description
Supelco
Ritalinic acid hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Ritalinic acid, 99% (CP)