MilliporeSigma
  • Usefulness of sweat testing for the detection of methylphenidate after fast- and extended-release drug administration: a pilot study.
Usefulness of sweat testing for the detection of methylphenidate after fast- and extended-release drug administration: a pilot study.
Therapeutic drug monitoring (2010-06-11)
Emilia Marchei, Magí Farré, Ricardo Pardo, Oscar Garcia-Algar, Manuela Pellegrini, Roberta Pacifici, Simona Pichini
ABSTRACT
The pharmacokinetics of methylphenidate (MPH), a prescription amphetamine derivative used in the treatment of attention-deficit hyperactivity disorder, has been amply described in conventional biological matrices. Recently, the excretion of MPH and its principal metabolite, ritalinic acid (RA) in oral fluid and plasma after a single drug administration has been described. The aim of this study was to describe the excretion of MPH and RA in sweat after the administration of a single dose of either fast-release or extended-release MPH. Three male subjects received 2 simultaneous oral doses of 10 mg fast-release MPH, and 1 male subject received one dose of 20 mg extended-release MPH. Sweat patches were applied to the back of each participant and removed at timed intervals. MPH and RA were determined in patches using a previously validated liquid chromatography-electrospray ionization mass spectrometric method. MPH was detected in sweat after the administration of fast- and extended-release formulations. For the fast-release formulation, MPH appeared in the sweat patches 2 hours after administration with a maximum of 15.9 nanogram per patch, reached after 24 hours. Mean total MPH excreted was 0.02 mg (about 0.08% of the administered dose). For the extended-release formulation, MPH appeared in the sweat 5 hours after administration and reached a maximum of 34.3 nanogram per patch after 24 hours. Mean total MPH excreted was 0.04 mg (about 0.18% of the administered dose). RA was not detected in either of the sweat patches probably because of its acidic properties. Measuring MPH in sweat patches can be a viable alternative to urine testing for noninvasive monitoring of use and misuse of the drug.
Sigma-Aldrich
Ritalinic acid, 99% (CP)
Supelco
Ritalinic acid hydrochloride solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®