MilliporeSigma
  • Chronic treatment with the neuroactive steroid ganaxolone in the rat induces anticonvulsant tolerance to diazepam but not to itself.
Chronic treatment with the neuroactive steroid ganaxolone in the rat induces anticonvulsant tolerance to diazepam but not to itself.
The Journal of pharmacology and experimental therapeutics (2000-11-18)
D S Reddy, M A Rogawski
ABSTRACT
Ganaxolone (3alpha-hydroxy-3beta-methyl-5alpha-pregnane-20-one), an orally active synthetic analog of the neuroactive steroid allopregnanolone, is a positive allosteric modulator of gamma-aminobutyric acid(A) receptors with anticonvulsant properties. We sought to determine whether tolerance occurs to the anticonvulsant activity of ganaxolone in the pentylenetetrazol seizure test and whether there is cross-tolerance with diazepam. Rats were treated with two daily injections of a 2 x ED(50) dose of ganaxolone (7 mg/kg s.c.), diazepam (4 mg/kg i.p.), or vehicle for 3 or 7 days. On the day after the chronic treatment periods, the anticonvulsant potencies of ganaxolone and diazepam were determined. The ED(50) values for ganaxolone after 3- and 7-day treatment with ganaxolone were not significantly different from that in naive rats (ED(50) = 3.5 mg/kg). In contrast, in animals that were treated chronically with ganaxolone for 7 days, there was a significant reduction in the anticonvulsant potency of diazepam (ED(50) = 4.0 versus 1.9 mg/kg for naive controls). Chronic treatment with diazepam was not associated with a reduction in the potency of ganaxolone, but there was a reduction in the potency of diazepam (ED(50) = 3.7 mg/kg). Plasma ganaxolone determinations indicated that the pharmacokinetic properties of ganaxolone were unchanged after 7-day chronic ganaxolone treatment. The estimated equilibrium plasma concentrations of ganaxolone associated with threshold (750-950 ng/ml) and 50% seizure protection (1215-1295 ng/ml) were similar in naive and chronically treated rats. We conclude that there is no tolerance to the anticonvulsant activity of ganaxolone nor is there cross-tolerance to ganaxolone when tolerance develops to diazepam. However, there is cross-tolerance to diazepam with chronic ganaxolone treatment.
Sigma-Aldrich
Ganaxolone, ≥98% (HPLC), solid