MilliporeSigma
  • Modification of excitation-contraction coupling by 4-chloro-m-cresol in voltage-clamped cut muscle fibres of the frog (R. pipiens).
Modification of excitation-contraction coupling by 4-chloro-m-cresol in voltage-clamped cut muscle fibres of the frog (R. pipiens).
The Journal of physiology (1999-02-02)
A Struk, W Melzer
ABSTRACT
1. The effect of 5 microM 4-chloro-m-cresol (4-CmC) on voltage-controlled Ca2+ release was studied in cut muscle fibres of the frog loaded with internal solutions containing 15 mM EGTA. Fibres were voltage clamped using a double Vaseline gap system, and Ca2+ signals were recorded with the fluorescent indicator dye fura-2 2. Resting intracellular free Ca2+ concentration increased from 61 to 100 nM upon application of 4-CmC. 3. Both peak rate of release of intracellularly stored Ca2+ and the steady level attained after 50 ms of depolarization increased, but the potentiation of the latter was more pronounced (by a factor of 1.7 versus 1.3). The voltage of half-maximal activation remained unchanged. 4. Non-linear intramembranous charge movements showed no significant change in voltage dependence while the maximal charge displaced by depolarization increased by 25 %. 5. The dependence of peak release flux on total intramembranous charge was not different in 4-CmC, but for the steady level of release the steepness of the relation increased by a factor of 1.3. 6. The stimulating effect of 5 microM 4-CmC on depolarization-induced Ca2+ release resembled the potentiation by 0.5 mM caffeine. However, 0.5 mM caffeine increased the peak and steady levels of the release rate by a similar factor and caused no increase in the resting free calcium concentration, indicating different modes of action of the two substances. 7. Neither 5 microM 4-CmC nor 0.5 mM caffeine led to a loss of voltage control of Ca2+ release during repolarization after short depolarizations, as has been reported previously for caffeine. Potentiated Ca2+ release could be terminated by repolarization as fast as under control conditions both with 15 mM and 0.1 mM internal EGTA. 8. The effects of 4-CmC may result from a direct opening of the release channel combined with an enhancement of the transduction mechanism that couples channel opening to displacement of voltage sensor charges.
Sigma-Aldrich
4-Chloro-2-methylphenol, 97%
Supelco
4-Chloro-2-methylphenol, PESTANAL®, analytical standard