MilliporeSigma
  • Inhibitory effect of 2,5-dimethylpyrazine on oxytocic agent-induced uterine hypercontraction of normal or pregnant female rats.
Inhibitory effect of 2,5-dimethylpyrazine on oxytocic agent-induced uterine hypercontraction of normal or pregnant female rats.
Biological & pharmaceutical bulletin (1998-06-23)
K Yamada, Y Kobayashi, H Fujihara, A Ohta
ABSTRACT
Intraperitoneal administration of 2,5-dimethylpyrazine (2,5-DMP) was found to inhibit oxytocin- and prostaglandin F2alpha-induced tetanic uterine contractions in normal or pregnant female rats. This suggests that 2,5-DMP may be used as a countercontraction agent or relaxant for preventing oxytocic agent-induced medical accident including uterine rupture or pressure death of the fetus due to uterine contractions.
Sigma-Aldrich
2,5-Dimethylpyrazine, ≥98%, FG
Sigma-Aldrich
2,5-Dimethylpyrazine, 98%