MilliporeSigma
  • Vibrational analysis of substituted benzonitriles. III. Transferability of force constants--the case of some halogeno-, methoxy- and nitro-benzonitriles.
Vibrational analysis of substituted benzonitriles. III. Transferability of force constants--the case of some halogeno-, methoxy- and nitro-benzonitriles.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (1998-01-23)
A P Kumar, G R Rao
ABSTRACT
A zero-order normal coordinate analysis of both the in-plane and out-of-plane vibrations was made for 2-chloro, 6-fluorobenzonitrile, s-trichlorobenzonitrile, p- and m-methoxybenzonitriles and m-nitrobenzonitrile, transferring the force constants from our earlier work. The observed and calculated frequencies agree with an average error of 16.8 cm-1, demonstrating the transferability of the force constants obtained previously. On the basis of calculated potential energy distributions and eigenvectors, several assignments suggested by earlier workers have been revised.
Sigma-Aldrich
3-Nitrobenzonitrile, 98%
Sigma-Aldrich
4-Nitrobenzonitrile, 97%
Sigma-Aldrich
4-Methoxybenzonitrile, 99%