MilliporeSigma
  • Determination of hydrogen peroxide with N,N-diethylaniline and 4-aminoantipyrine by use of an anion-exchange resin modified with manganese-tetrakis(sulphophenyl)porphine, as a substitute for peroxidase.
Determination of hydrogen peroxide with N,N-diethylaniline and 4-aminoantipyrine by use of an anion-exchange resin modified with manganese-tetrakis(sulphophenyl)porphine, as a substitute for peroxidase.
Talanta (1987-07-01)
Y Saito, M Mifune, S Nakashima, J Odo, Y Tanaka, M Chikuma, H Tanaka
ABSTRACT
Amberlite IRA 900 anion-exchange resin modified with manganese-tetrakis(sulphophenyl)-porphine has been used as a catalyst instead of peroxidase for the determination of hydrogen peroxide by the reaction 2H(2)O(2) + N,N-diethylaniline + 4-aminoantipyrine (catalyst)--> quinonoid dye (lambda(max) 550 nm) + 4H(2)O. The apparent molar absorptivity for hydrogen peroxide was 1.1 x 10(4) 1.mole(-1).cm(-1), coefficient of variation 0.7%. This value is approximately 84% of that obtained by the use of peroxidase as catalyst. Similar conditions to those in the enzymatic reaction were suitable for use of the modified resin as catalyst, and the results show it to be a good substitute for peroxidase in this reaction system.
Product Description
Supelco
4-Aminoantipyrine, for spectrophotometric det. of H2O2 and phenols, ≥98.0%