MilliporeSigma
  • Plasma total odd-chain fatty acids in the monitoring of disorders of propionate, methylmalonate and biotin metabolism.
Plasma total odd-chain fatty acids in the monitoring of disorders of propionate, methylmalonate and biotin metabolism.
Journal of inherited metabolic disease (1996-01-01)
M Coker, J B de Klerk, B T Poll-The, J G Huijmans, M Duran
ABSTRACT
Total plasma odd-numbered long-chain fatty acids were analysed in patients with methylmalonic acidaemia (vitamin B12-responsive and unresponsive), combined methylmalonic acidaemia/homocystinuria (CblC), propionic acidaemia (both neonatal-onset and late-onset), biotinidase deficiency and holocarboxylase synthase deficiency, as well as in hospital controls. Total odd-numbered long-chain fatty acids (C15:0, C17:1 and C17:0) were expressed as a percentage of total C12-C20 fatty acids. Control values were 0.72% +/- 0.31% (n = 12). Normalization of the percentage of odd-chain fatty acids occurred in all vitamin-responsive patients, following the institution of vitamin treatment. In general the neonatal-onset propionic acidaemia and B12-unresponsive methylmalonic acidaemia patients had the highest plasma odd-chain fatty acid concentrations, which correlated with the clinical condition but not with the urinary excretion of methylcitrate or methylmalonate. Plasma odd-chain fatty acid concentrations and methylmalonate excretions in CblC patients reacted very well to vitamin B12 treatment, but with no clinical response. Measurement of plasma odd-chain fatty acids is of no value for the monitoring of defects of biotin metabolism.
Sigma-Aldrich
Dimethyl malonate, purum, ≥96.0% (GC)
Sigma-Aldrich
Dimethyl malonate, 98%