MilliporeSigma
  • Determination of acylcarnitines in urine of patients with inborn errors of metabolism using high-performance liquid chromatography after derivatization with 4'-bromophenacylbromide.
Determination of acylcarnitines in urine of patients with inborn errors of metabolism using high-performance liquid chromatography after derivatization with 4'-bromophenacylbromide.
Clinica chimica acta; international journal of clinical chemistry (1993-07-16)
B J Poorthuis, T Jille-Vlcková, W Onkenhout
ABSTRACT
A high-performance liquid chromatographic method is presented for the determination of urinary acylcarnitines. After solid phase extraction on silica columns the acylcarnitines are converted to 4'-bromophenacyl esters with 4'-bromophenacylbromide in the presence of N,N-diisopropylethylamine. Complete derivatization was achieved at 37 degrees C within 30 min. The 4'-bromophenacyl esters were separated by high-performance liquid chromatography on a Hypersil BDS C8 reversed-phase column with a binary gradient containing varying proportions of acetonitrile, water and 0.1 M triethylamine phosphate buffer. Essentially baseline separation was obtained with a standard mixture containing 4'-bromophenacyl esters of carnitine and synthetic acylcarnitines of increasing chain length ranging from acetyl- to palmitoylcarnitine. The method was used to obtain urinary acylcarnitine profiles from patients with propionic, methylmalonic and isovaleric acidemia and with medium-chain and multiple acyl-CoA dehydrogenase deficiency. Quantification of the acylcarnitines was achieved using undecanoylcarnitine as internal standard.
Sigma-Aldrich
N-Ethyldiisopropylamine, BASF quality, ≥98.0%
Sigma-Aldrich
N-Ethyldiisopropylamine solution, suitable for peptide synthesis, ~2 M in 1-methyl-2-pyrrolidinone
Sigma-Aldrich
N,N-Diisopropylethylamine, 99.5%, biotech. grade
Sigma-Aldrich
N,N-Diisopropylethylamine, ReagentPlus®, ≥99%
Sigma-Aldrich
N,N-Diisopropylethylamine, purified by redistillation, 99.5%