MilliporeSigma
  • Genetic toxicological of some plant flavonoids by the micronucleus test.
Genetic toxicological of some plant flavonoids by the micronucleus test.
Mutation research (1981-05-01)
R K Sahu, R Basu, A Sharma
ABSTRACT
4 important flavonoid compounds, namely quercetin, kaempferol, neohesperidin dihydrochalcone and rutin, were administered to male mice for the detection of gross chromosomal anomalies by the micronucleus test. The first 3 compounds were positively clastogenic to different extents, whereas the 4th was negative.
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥95% (HPLC)
Sigma-Aldrich
Neohesperidin dihydrochalcone, ≥96%, FG
Supelco
Neohesperidin dihydrochalcone, analytical standard