MilliporeSigma
  • Characterization of sialoglycoproteins of rat epididymal fluid and spermatozoa by periodate-tritiated borohydride.
Characterization of sialoglycoproteins of rat epididymal fluid and spermatozoa by periodate-tritiated borohydride.
Journal of reproduction and fertility (1983-01-01)
P Toowicharanont, M Chulavatnatol
ABSTRACT
Sialoglycoproteins of rat epididymal fluid and spermatozoa were radiolabelled by the NaIO4/KB3H4 method. At least 10 sialoglycoproteins of the epididymal fluids could be consistently demonstrated by polyacrylamide gel electrophoresis in sodium dodecyl sulphate. Two major ones (Mr 21000 and 66000) were present in the fluids of the caput and cauda epididymidis. Two (Mr 28000 and 40000) were found only in the former and two (Mr 32000 and 42000) only in the latter. There were at least 11 sialoglycoproteins bound to the epididymal spermatozoa. During epididymal transport, 8 sialoglycoproteins on the spermatozoa decreased, one (Mr 48000) remained constant and one (Mr 31000) increased in amount. During sperm maturation, some sperm-bound sialoglycoproteins, especially 3 of small molecular weights, became more resistant to the treatments with neuraminidase, trypsin and Triton X-100.
Sigma-Aldrich
Potassium borohydride, purum, ≥97.0% (RT)
Sigma-Aldrich
Potassium borohydride, ≥97%
Sigma-Aldrich
Potassium borohydride, 99.9% trace metals basis