MilliporeSigma
  • Guinea pig maximization test: effect of type of Freund's complete adjuvant emulsion and of challenge site location.
Guinea pig maximization test: effect of type of Freund's complete adjuvant emulsion and of challenge site location.
Dermatosen in Beruf und Umwelt. Occupation and environment (1985-01-01)
K E Andersen
ABSTRACT
Guinea pig maximization tests (GPMT) with chlorocresol were performed to ascertain whether the sensitization rate was affected by minor changes in the Freund's complete adjuvant (FCA) emulsion used. Three types of emulsion were evaluated: the oil phase was mixed with propylene glycol, saline with 30% (v/v) ethanol or saline, respectively. Relative viscosity was used as one measure of physical properties of the emulsion. Higher degrees of sensitization (but not rates) were obtained at the 48 h challenge reading with the oil/propylene glycol and oil/saline + ethanol emulsions compared to the saline/oil emulsion. Placing of the challenge patches affected the response, as simultaneous chlorocresol challenge on the flank located 2 cm closer to the abdomen than the usual challenge site gave decreased reactions.
Sigma-Aldrich
4-Chloro-2-methylphenol, 97%
Supelco
4-Chloro-2-methylphenol, PESTANAL®, analytical standard